MS Series High Capacity Balances, Mettler-Toledo
Proveedor: Mettler-Toledo
NewClassic
11145016 11145141 11145013 11145141 11145028 11145028 11145013 11145016
97035-650EA 213210 MXN
CA97035-656 97035-656 97035-654 97035-652 97035-650 CA97035-650 CA97035-652 CA97035-654
MS Series High Capacity Balances, Mettler-Toledo
Balanzas
The NewClassic MS series top-loading, high-capacity balances offer quality in an economical package.
  • Large Capacity Weighing Pan for Comfortable Weighing of Bulky Items
  • Overload Protection Guards the Weighing Cell Against Excess Weight
  • Chemical-Resistant, Die-Cast Aluminum Housing
These fast, reliable balances provide quick, accurate results in a variety of routine applications.
The MS series offers six models with capacities ranging from 12.2 to 32.2kg and readabilities ranging from 0.1 to 1g. Balances are equipped with the rugged, high-performance MonoBloc weighing cell, a large weighing platform, and an HCD (high contrast display) with large numerals, clear symbols, and an intuitive menu. Built-in applications include piece counting, percent weighing, and dynamic weighing. An RS-232 interface (changeable to LocalCAN™ universal interface) permits easy communication exchange. All balances come with full taring range.
Certificaciones: METTLER TOLEDO® provides ISO 9001 manufacturing and nationwide ISO 9001 field service. UL listed and CSA certified.
Información para pedidos: Balances are supplied with an in-use cover and a one-year manufacturer's warranty. Extended on-site warranty covers all costs associated with repair during the second and third years. To order optional METTLER TOLEDO® calibration weights, contact your Avantor sales representative. The optional Gold and Silver calibration packages are offered to assure continual reliability and maintain optimum balance performance. Gold service packages are suitable for many regulated and unregulated balance uses requiring documentation to support installation, including a NIST traceable calibration certificate. Silver service packages are suitable for many unregulated balance uses, requiring no significant documentation to support installation. Contact your Avantor representative for package specifics.