Guardian™ Airflow Monitor Kits, Labconco®
Proveedor: Labconco
Guardian™
3908801 9743201 9743203 9743207 9743207 9743210 3908801 9743201 9743208 9743209 3908800 9743205 9743204
46620-470EA 66161.39 MXN
14222-630 82007-286 46620-470 82010-706 82010-708 470203-272 82001-232 470203-276 470203-190 82007-280 74920-162 82007-284 82007-282
Guardian™ Airflow Monitor Kits, Labconco®
Vitrinas de gases y recintos Campanas de gases químicos
Guardian™ airflow monitors continuously check the face velocity of laboratory hoods and enclosures and alert the operator to airflows that drop outside preset levels with audible and visual alarms. The audible alarm can be muted at the touch of a switch.
The Guardian™ digital monitor displays actual face velocity on an LCD screen and includes a programmable switch to open the hood damper
The right fixture panel is factory-prepared for an airflow monitor on the following nonexplosion-proof laboratory hoods: Protector Premier™, Protector XStream®, Protector XL™, Protector Stainless Steel, Protector PVC, Basic™, and Fiberglass 30 hoods. On the ClassMate® hoods, the monitor can be installed in the front panel of the hood. On the Protector® Demonstration and XVS hoods, and the Protector® Workstation, the monitor can be installed on the top of the hood.
The Guardian™ 1000 digital monitor features an LCD of actual airflow in fpm or m/sec. from 0–999fpm. On the Guardian™ Jr. monitor, a green light glows when the velocity is normal. If the face velocity falls below setpoint for more than six seconds, the audible alarm sounds and the alert indicator glows red.
Información para pedidos: All items supplied with face plate with circuit board, electrical power pack, vinyl tubing, wiring instructions, and epoxy-coated steel casing when needed.
Información de suministro: Items indicated below as REDISHIP units can be shipped within 48 hours of receiving your order.