HEPA Filter Packs, Labconco®
Proveedor: Labconco
2240100 2240000 2244200
30181-651EA 105501.85 MXN
30181-651 30181-655 27682-152
HEPA Filter Packs, Labconco®
Vitrinas de gases y recintos Campanas de gases químicos
This high-efficiency, particulate air filter is for use in non-radioactive particulate applications
Rated for 1000cfm airflow with initial 1in.H2O static pressure drip, the filters have a replaceable HEPA filter media that removes 99.97% of all particles 0.34µm or greater. Filter packs are furnished with a clamping frame and duct connections.