Specifically designed to operate with MicroFunnel filter funnels when performing the MF technique for microbial analysis.
- Designed to operate with MicroFunnels
- Efficient operation
- No adapters required
The simple, efficient operation is conducted by placing filter funnels directly onto the manifold, adding liquid, and turning on the vacuum. No adapters or rubber stoppers are required to hold filter funnels in place. The manifold body is constructed of anodized aluminum. Autoclavable at 121 to 123°C (250 to 253°F), 1.0bar (15psi) for 15–20 minutes.
Three-place and six-place manifolds are capable of processing multiple samples simultaneously. Valves are manufactured from chrome-plated brass. Valve O-rings are made of Viton® rubber. Drain plugs are constructed of stainless steel, and feature 6.4mm (1/4") MNPT outlets. These materials make the unit easy to clean and compatible with a wide variety of chemicals. Hose barb adapter: 6.4mm (1/4") nylon.
One-place manifold has a stainless steel hose barb adapter: 6.4mm (1/4") straight, 3.2mm (1/8") MNPT.