Whatman™ Filter Funnel Plates, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
1950-114 1950-117 1950-109 1950-109 1950-117 1950-114
80085-990EA 4500 MXN
CA80085-990 80085-992 80085-990 CA80085-992 28200-753 CA28200-753
Whatman™ Filter Funnel Plates, Whatman products (Cytiva)
Embudos Embudos de filtración de membrana
Designed for use with Whatman glass microfiber filters
Three-piece construction permits quick filter changing and easy cleaning. Reservoir, base and stem are borosilicate glass with ground contact flanges. Clamping pressure provides a leak-tight seal. Solutions pass through filter and all retained solids are deposited within filter circle. Clamps are plastic-coated for chemical resistance. Replaceable filter support plates of transparent acrylic are suitable for most aqueous solutions and some dilute acids at working temperatures to 65°C maximum. Optional polypropylene plate 28200-753 (9cm) is available.
Ideal for water testing and for biomedical, microbiological and clinical chemistry.
Información para pedidos: Supplied with acrylic filter support plate and three clamps.