VWR® Quantitative Filter Paper, Grade 494
Proveedor: Avantor
VWR®
28297-930 28307-134
28307-134PK 509.72 MXN
28297-930 28307-134
VWR® Quantitative Filter Paper, Grade 494
Filtros Papeles de filtros Papel de filtro cuantitativo
Useful for very fine precipitates such as barium and lead sulfates, stannic and nickel sulfides, and calcium hydroxide.