Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.01%), Grade 589/3, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
10300263 10300210 10300214 10300263 10300211 10300212 10300213 10300211 10300214 10300210 10300213 10300212
14224-450PK 2500 MXN
14224-450 CA76423-780 89233-654 76423-780 CA89233-654 CA14224-450 CA14224-444 14224-444 76423-778 10034-630 CA76423-778 CA11012-658
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.01%), Grade 589/3, Whatman products (Cytiva)
Filtros Papeles de filtros Papel de filtro cuantitativo
Whatman Grade 589/3 ashless filter paper from Cytiva is particularly useful in capturing very fine particulate samples such as insoluble molecules in fats or oils or supporting ash free dry mass.
  • High purity (<0.01% ash) for ash free dry mass methods
  • Blue ribbon filter
  • Very fine precipitate retention (class 2d acc. to DIN 53 135)
  • Herzberg filtration speed of 750 s*
  • Also available prepleated as Grade 589/3 ½
*The time taken to filter 100 ml water (20° C) through a filter area of 10 cm² at a constant pressure of 5 cm water column.
Grade 589/3 filter paper from Cytiva's business supports accurate measurement of ash free dry mass. The blue ribbon filter is an established component in a variety of quantitative analysis methods, such as quantification of insoluble contaminants in animal and vegetable fats and oils. Ash free dry mass.