Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 542, Whatman Products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
1542-055 1542-150 1542-055 1542-090 1542-240 1542-400 1542-110 1542-400 1542-125 1542-240 1542-150 1542-070 1542-110 1542-125 1542-090 1542-185 1542-185 1542-070
28297-400EA 140.63 MXN
CA10147-892 10147-894 10147-892 CA11013-102 CA10147-894 28297-400 CA28297-398 CA28297-400 10035-284 28297-390 CA28297-390 CA28297-392 CA28297-394 CA28297-396 28297-398 28297-396 28297-394 28297-392
Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 542, Whatman Products (Cytiva)
Filtros Papeles de filtros Papel de filtro cuantitativo
Cytiva's Whatman Grade 542 filters are acid hardened to allow high retention of fine precipitates in applications such as metal determinations and hydroxide collection.
  • High wet-strength
  • Acid-hardened for resistance to strong acid and alkali solutions
  • Nominal particle retention rating of 2.7 µm
  • Herzberg filtration speed of 2510 s
  • Extremely low ash content (0.006% ash maximum)
Along with Grade 542 cellulose filter paper, the Whatman range of hardened quantitative papers from Cytiva's business includes fast Grade 541 and general-purpose Grade 540 filters. All three grades are designed for use in quantitative analysis with strong acid/alkali solutions.
Hardened Grade 542 quantitative papers enable high retention of fine particles in chemically harsh conditions. When acting as a gravimetric filter, this cellulose filter paper is therefore well suited for use in capturing fine precipitates in applications such as metal determinations and hydroxide collection.