Whatman™ Grade 595 Qualitative Filter Papers, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
10311611 10311610 10311612 10311610 10311687 10311687 10311611 10311612
10034-642PK 800 MXN
CA11012-616 10035-780 10034-662 CA11012-670 CA11012-882 10034-642 28430-109 CA28430-109
Whatman™ Grade 595 Qualitative Filter Papers, Whatman products (Cytiva)
Filtros Papeles de filtros Papel de filtro cualitativo
A standard grade filter used for many routine analytical applications in different industries (e.g., particle separation from food extracts or filtration of solids from digested environmental samples for iCP/AAs analysis).
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Grade 595: 4 - 7 µm
Very popular, thin filter paper, medium-fast with medium to fine particle retention. Used for many routine analytical applications in different industries, (e.g., particle separation from food extracts or filtration of solids from digested environmental samples, e.g., for ICP/AAS analysis).