VWR® Horizontal Centrifuges
Proveedor: VWR
VWR®
00-045-037-000 00-018-037-000
95037-282EA 186097.15 MXN
95037-278 95037-282
VWR® Horizontal Centrifuges
Centrífugas Centrífugas de sobremesa
Welded steel centrifuges are capable of separating platelet-rich, platelet-poor, and platelet-free plasma.
  • Platelet separation
  • Fast processing
  • Rotor, tube holders, and caps included
  • Safety lock lid
All centrifuges feature a patented horizontal rotor design for faster loading and processing; a clear safety lock lid with a mechanical latch and indicator lights; and a brushless, double-encased motor for quiet, low-vibration operation.
Single-speed centrifuge 95037-278 features a digital timer with memory recall. Variable speed centrifuges 95037-280 and 95037-282 feature an electronic timer with display and memory for ten settings, variable acceleration and deceleration with electronic brake, electronic variable speed control (500rpm to maximum rpm in increments of 50) with display, an audible end-of-run indicator, and out-of-balance detection in which centrifuge will come to a safe, controlled stop if an imbalance is sensed.
Certificaciones: UL listed. CSA certified.
Información para pedidos: All centrifuges include rotor and tube holders.