Acrodisc® Syringe Filters for Prefiltration, Cytiva (Formerly Pall Lab)
Proveedor: Cytiva
Acrodisc®
4529 4488 AP-4523 4189 AP-4529 AP-4000 AP-4002 4489 4489 AP-4000 4459 4488 4568 4459 4524 AP-4529 4487 AP-4527 AP-4189 AP-4001 AP-4189 4529 AP-4002 AP-4568 AP-4523 4189 4568 AP-4527 AP-4001 AP-4568 4473 4523 4523 4524 4473 4487
28143-960CS 40307.18 MXN
28143-960 28143-962 28143-963 28143-986 28143-987 CA34181-076 CA28143-998 CA28143-954 CA28143-975 CA16003-732 28143-904 CA16003-734 CA28143-959 28143-907 CA16003-736 28143-908 CA28143-908 28143-380 CA28143-394 CA28143-990 34181-092 CA28143-963 28143-975 28143-998 CA28143-962 28143-954 CA28143-987 16003-732 CA28143-986 16003-734 16003-736 34181-076 CA28143-904 CA28143-907 CA28143-380 28143-394
Acrodisc® Syringe Filters for Prefiltration, Cytiva (Formerly Pall Lab)
Filtros Filtros de jeringas
For prefiltering heavily contaminated aqueous or solvent based samples.
  • Increased pore size for reduced clogging
  • Maximized throughput
  • Wide range of pore sizes available
The glass fiber membrane has very low extractables. Filters are made of 100% binder-free borosilicate glass fiber.
The Versapor® (acrylic copolymer) membrane filters feature large pore sizes ideal for prefiltration. These filters are color-coded by pore size for easy identification.
Polypropylene housing, female Luer-lock inlet and male slip Luer outlet.
Certificaciones: Acrodisc PSF syringe filters are automation certified to assure smooth operation and worry-free performance 24 hours a day in automated workstations.
Envase: Syringe filters packed in cases of 200 are available in four packs of 50 or in an AutoPack™ format. The AutoPack™ format consists of eight tubes with 25 syringe filters stacked on top of each other, which is specifically designed for the Caliper™ robotic workstations.