Target2™ Nylon Syringe Filters, Thermo Scientific
Proveedor: Thermo Scientific
Target®
F2504-1 F2513-1 F2504-2 F2500-1 F2500-50 F2513-2 F2502-1 F2500-2 F2502-2 F2500-12
66030-859EA 6015.88 MXN
66064-395 66030-839 66030-859 66030-837 66030-849 66030-872 66030-861 66030-870 66064-847 66030-851
Target2™ Nylon Syringe Filters, Thermo Scientific
Filtros Filtros de jeringas
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
  • Hot-tested for physical properties and UV extractables
Available in sizes and membrane types to meet any application need, Thermo Scientific™ Target2™ syringe Filters provide cleaner sample extracts by removing interfering materials and fine particles down to the rated pore size. Target2 Nylon Syringe Filters are a good choice for most general laboratory filtration, as well as filtration of most HPLC samples.