Target® Syringe Filters, Cellulose Acetate, National Scientific™
Proveedor: National Scientific
Target®
F2500-16 F2504-16 F2504-15
66064-854CS 40142.96 MXN
66064-854 66064-834 66064-836
Target® Syringe Filters, Cellulose Acetate, National Scientific™
Filtros Filtros de jeringas
Cellulose acetate membrane for biological or protein-based samples.
  • Low nonspecific binding with limited chemical resistance
  • Good flow rate characteristics
Use the 4 mm filters for limited volume samples up to 2 ml, and the 13 and 25 mm filters for volumes up to 100 ml. The 0.2 µm filters can be used for 'cold' sterilization.