VWR® Syringe Filters
Proveedor: VWR
VWR®
28145-489 28145-500 28145-495 28145-501 28145-493 28145-503 28145-501 28145-489 28145-475 28145-477 28145-493 28145-479 28145-495 28145-483 28145-499 28145-491 28145-485 28145-477 28145-491 28145-487 28145-505 28145-497 28145-481
28145-481CS 1797.05 MXN
CA28145-491 28145-481 28145-493 CA28145-493 28145-483 28145-495 28145-491 89041-306 28145-505 28145-489 CA28145-477 28145-501 28145-479 CA28145-501 CA28145-489 28145-503 CA28145-495 28145-485 28145-475 28145-497 28145-487 28145-499 28145-477
VWR® Syringe Filters
Filtros Filtros de jeringas
Polypropylene, nylon, and PTFE membranes are ideal for sample preparation and small volume chemical filtration.
  • Ideal for sterile filtration
  • Manufacturing process certified to ISO 9001
  • Biosafe according to Class VI plastics tests
Syringe filters with cellulose acetate or polyethersulfone membranes are ideal for sterile filtration of buffers, tissue culture media or additives, and other biologicals. The polyethersulfone membrane offers high flow rates. Nonsterile syringe filters are excellent for HPLC applications. Each filter has a female Luer-lock inlet and a male slip Luer outlet.
Información para pedidos: Nylon membrane filters 89041-306 are bulk packed in bags of 1000, four bags to a case.
Precaución: Note: PTFE membrane is hydrophobic and not intended for use with aqueous solutions.