Eppendorf® Microcentrifuges, Models 5430 and 5430R
Proveedor: Eppendorf
022620557 022620623 022620667 022620656 022620612 022620601 022620690 022620584 022620634 022620645 022620572 022620638 022620601 022620562 022620509 022620623 2231000769 022620642 022620568 022620557 022620541 022620513 022620634 022620666 022620511 022620689 022620541 022620580 022620562 2231000770 022620690 022620540 022620560 022620596 022620642 022620533 022620540 022620638 022620678 022620596 022620645 022620625 022620584 022620572 022620603 022620663 022620568 022620533 022620659 022620667 022620678 022620509 022620560 022620576 022620511 022620656 022620612 022620665
490015-774EA 255856.85 MXN
CA10030-958 CA10030-956 CA10030-996 97027-862 76460-824 97027-860 76460-822 89422-672 97027-858 CA97027-866 97027-856 97027-854 CA97027-862 490015-774 89422-676 CA80094-172 97027-852 CA80094-178 CA97027-860 CA80094-176 97027-850 CA97027-858 97027-848 CA97027-856 97027-846 CA97027-854 97027-844 CA97027-852 97027-842 80094-172 CA80094-182 CA80094-180 80094-176 CA97027-850 97027-840 10030-960 80094-178 97027-838 CA97027-846 75875-088 CA97027-844 CA97027-842 CA97027-840 75875-084 CA97027-848 10030-958 80094-182 10030-956 CA10030-960 10030-996 97027-866 75875-650 97027-864 75875-090 80094-180 75875-092 CA97027-838 75875-094
Eppendorf® Microcentrifuges, Models 5430 and 5430R
Centrífugas Microcentrífugas
The unique cross-over microcentrifuges 5430 and 5430 R combine the features of a microcentrifuge (small footprint) and multipurpose centrifuge (versatility) in one instrument.
  • Remarkable versatility with 12 different available rotors
  • Soft-touch one-finger lid closure for ergonomic operation
  • Menu-driven, multi lingual operation menu (English, German, French, Spanish)
  • Large backlit display
  • 5 program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Automatic imbalance detection
  • Eppendorf QuickLock® System
These centrifuges spin rotors for Eppendorf tubes and PCR strips as you would expect from any microcentrifuge. But additionally, in a compact size, centrifuges 5430 and 5430 R also accommodate rotors for microplates and 15/50 mL conical tubes. Now, the rotor program has been expanded with additional Eppendorf QuickLock® rotors. This includes 48-place rotors (1.5/2.0 mL), a 16-place rotor for Eppendorf Tubes® 5.0 mL and a swing-bucket rotor (24 x 1.5/2.0 mL). Centrifuges also feature Eppendorf QuickLock® System for quickly opening and closing the rotor lid.