Safe-Grind® Plastic-Coated Tissue Grinders with Fluoropolymer Resin Pestles, Wheaton
Proveedor: DWK Life Sciences
Safe-Grind®
358005 358009 358007 358003 358011
62400-314CS 8660.9 MXN
62400-314 62400-336 62400-325 62400-303 62400-347
Safe-Grind® Plastic-Coated Tissue Grinders with Fluoropolymer Resin Pestles, Wheaton
Homogenizadores Trituradoras tisulares
For sure grip and safe grinding of homogenate even with power homogenization.
Heavy, clear plastic coating on Potter-Elvehjem style tissue grinders protects mortar from scratches, and provides a surer grip. Coating also helps contain glass and homogenate in the event of a breakage. Transparent coating allows unobstructed view of homogenate. Autoclavable.