Elbows, Labconco
Proveedor: Labconco
7027300 5605100 7027400 5605200 4719100 5602100 4708800 4708700 4719000
30182-136EA 21408 MXN
82006-930 82006-932 30182-144 30182-111 30182-136 30152-640 30182-124 30182-326 30182-290
Elbows, Labconco
Ducting Supplies
PVC elbows, both 45° and 90°, are compatible with thermoplastic duct
Designed and engineered for quick installation and minimum pressure losses, elbows feature belled end connections to receive PVC duct directly.