Heidolph® MR Hei-Tec Magnetic Hotplate Stirrers
Proveedor: Heidolph
Heidolph®
036110550CA 036110550
89029-830EA 34350.04 MXN
CA11009-526 89029-830
Heidolph® MR Hei-Tec Magnetic Hotplate Stirrers
Imánes de agitación Agitadores magnéticos con placas calefactoras
Hotplate stirrers feature excellent heat conduction and distribution as well as resistance against scratches and chemicals.
Round hotplate is constructed of Silumin aluminum alloy with a ceramic coating.
Información para pedidos: For heating baths, sensors, spin bars, support rods, and other accessories, contact your Avantor representative.