Revco® Digital Temperature Monitors, Thermo Scientific
Proveedor: Thermo Scientific
Revco®
6899 6900 6898
14228-506EA 34858.64 MXN
14228-502 14228-506 14228-504
Revco® Digital Temperature Monitors, Thermo Scientific
Registradores de datos
These digital temperature and power monitor units provide both alarm/monitoring and digital temperature display in a single, self-contained device.
  • An audible and visual warning is activated in case of power failure or overtemperature condition
  • Monitors can be installed on or adjacent to any freezer
One channel monitors feature a Velcro closure, 2 or 3 channel monitors feature a mounting bracket. Standard features include high alarm with mute and chirp, audio delay timer, remote alarm contacts (SPDT), LCD numeric display, NiCad battery backup, low voltage AC power supply (120 V), and 32L×6D mm (1¹/₄×¹/₄") temperature probe. The 2 or 3 channel monitors are supplied with a high and low alarm, and 8-character backlit adjustable LCD alphanumeric display.