Order Entry
Nalgene® Vacuum Chambers, Thermo Scientific
undefined
Nalgene® Vacuum Chambers, Thermo Scientific
  •  
    Material
    Capacity
    Color
    Ext.Ø
    Height
    Supplier No.
  • Vacuum Chamber with Vacuum Plate O.D. 19 cm (71/2")
    Vacuum Chamber with Vacuum Plate O.D. 19 cm (71/2")
    54929-051
    5305-0609
    Polycarbonate (Jar)
    Polypropylene (Plate)
    4.7 L
    Clear
    Jar: 16.8 cm (65/8")
    Plate: 19 cm (71/2")
    23.8 cm (9⅜")
    5305-0609
    Each (1UN)
    Each (1UN) - $7,525.73
    $7,525.73
    Case de 2 (1 X 1UN)
    Case de 2 (1 X 1UN) - $8,203.05
    $8,203.05
    Each (1UN)
       $7,525.73
    Case de 2 (1 X 1UN)
       $8,203.05
    Accessories
    Specifications
    Environmentally Preferable
  • Vacuum Chamber with Vacuum Plate O.D. 33 cm (13")
    Vacuum Chamber with Vacuum Plate O.D. 33 cm (13")
    54929-084
    5305-1212
    Polyetherimide (Jar)
    Polycarbonate (Plate)
    18.9 L
    Amber
    Jar: 30.5 cm (12")
    Plate: 33 cm (13")
    30.5 cm (12")
    5305-1212
    Case (1UN)
    Case (1UN) - $28,612.72
    $28,612.72
    Case (1UN)
       $28,612.72
    Accessories