VWR® Check Valves
Proveedor: VWR
VWR®
3296 3298 3295 3297
89094-714PK 1205.57 MXN
89094-708 89094-712 89094-714 89094-710
VWR® Check Valves
Válvulas Válvulas de vacío
Barbed check valves prevent damage to equipment, pumps, and flow systems by preventing backflow.
  • Chemical-resistant resin body and valve
  • Inert, high-purity fitting
  • Autoclavable
Chemical-resistant Kynar® resin body and Viton® resin valve ensure an inert, high-purity fitting that provides a smooth and clear flow path. Can be used with liquids and gases in chemical processing, pressure, and vacuum applications. Suitable for use with acids, alcohols, bases, and water.