Nalgene® Filter Holder with Receiver, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
300-4100 300-4050 300-4000
28199-450CS 35063.77 MXN
28199-450 28199-440 28199-406
Nalgene® Filter Holder with Receiver, Thermo Scientific
Portafiltros
Made of transparent polysulfone. Break-resistant and withstands repeated autoclaving (before autoclaving, set cover on filter unit without engaging threads).
Chemically resistant to bases, weak acids, and all aqueous solutions. Three cover ports allow venting and aseptic addition of samples. Receiver is graduated and has two side arms and two TPE caps.
Membrane for filtering liquids by either vacuum or pressure. The one-piece support plate provides maximum flow rate and high throughput. White, two-piece plate keeps the membrane flat for analysis of particulates or biological contaminants trapped on the surface. Clear, one-piece plate provides maximum flow rate and throughput for sterilization.
Información de suministro: Includes tubing adapter, which can be plugged with cotton for venting, and a receiver cover for conveniently storing sterile filtrate.
Precaución: For lab use only. Not for in vitro diagnosis or parenterals.