Whatman™ EPM 2000 Air Sampling Filter, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
1882-866 1882-047
28495-330EA 60.19 MXN
28495-420 28495-330
Whatman™ EPM 2000 Air Sampling Filter, Whatman products (Cytiva)
Filtros Filtros de fibras de vidrio
Cytiva's Whatman EPM 2000 air filters are used in high-volume PM-10 sampling equipment.
  • Air retention efficiency of 99.95 percent of 0.3 µm size particulates
  • Selected by the U.S. Environmental Protection Agency (EPA) for use in a nationwide network of HiVol air samplers
  • Manufactured according to ISO9001:2008 QMS standards
  • Available in both sheet and circle design
  • Sheets are individually numbered to facilitate identification
  • Binder-free
EPM 2000 from Cytiva's business is available as 100-piece packs of either 47 mm filter paper circles or 8 10" sheets.
Manufactured from 100% pure borosilicate glass, they enable detailed chemical analysis of trace pollutants with minimal interference.