Filling Machine Capsules, Cytiva (Formerly Pall Lab)
Proveedor: Cytiva
12121
CA28145-786
Filling Machine Capsules, Cytiva (Formerly Pall Lab)
Filtros Cápsulas de filtro de membrana
Ensure ultraclean products and accurate fills with pharmaceutical filling machines.
  • Connected upstream
  • Removes contaminants
  • Built-in air vent
Connected upstream of the filling machine nozzle, capsules filter out contaminants just before the product is packaged. Built-in air vent ensures accurate fills and uniform flow. Capsules are available with HT Tuffryn® (polysulfone) or Versapor® (acrylic copolymer) mebrane. Effective filtration area: 300 cm². Maximum operating pressure: 3.4 bar. Autoclavable. Capsules pass USP Class VI 121 °C Plastics Test.