Whatman™ Polycap™ TF Disposable Filter Capsules, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™ Polycap™
2602S 2702M 2702T 6700-3602 2700T 6700-7504 2603T 6700-7502 6710-7502 6710-7502 2703T 6710-3602 6711-3604 6700-7504 6700-3602 2702M 2804T 2803T 6701-7510 2802T 6700-7501 2702T 2802T 6711-3601 2801 6710-3602 2804T 2703T 2700T 6711-3602 6711-3601 6701-7510 2803T 6700-7502 6711-3604 2603T 6711-7502 6711-7502 2801 2800T 2601T 6700-3610 2601T 6700-7501 2800T 2602S 6700-3610 6711-3602
28137-874EA 5100 MXN
CA28137-880 CA11013-712 28137-872 CA28137-882 28137-874 10035-406 10147-952 10035-408 97059-550 CA11013-098 CA11013-230 CA11013-670 28137-716 CA11012-854 CA28137-720 28137-876 28137-878 CA28137-722 CA89013-354 10035-372 10035-374 10035-352 CA28137-716 28137-708 10035-370 CA11013-708 CA10147-952 10035-354 10035-410 10035-356 10035-412 28137-882 CA28137-872 28137-720 28137-880 CA11013-100 CA28137-878 CA97059-550 28137-728 CA11012-840 28137-722 CA28137-874 CA28137-876 89013-354 CA11012-838 CA28137-728 CA28137-708 CA11013-718
Whatman™ Polycap™ TF Disposable Filter Capsules, Whatman products (Cytiva)
Filtros Cápsulas de filtro de membrana
Cytiva's Whatman Polycap TF filters are made with durable, hydrophobic polytetrafluoroethylene (PTFE) membranes in a polypropylene housing for use with organic solvents and chemically aggressive solutions.
  • PTFE filter membrane is resistant to most solvents
  • Available in 0.05, 0.1, 0.2, 0.45, and 1.0 µm pore sizes
  • 0.05 µm pore size capsules capsules designed for ultra-clean applications
  • 1.0 µm pore size capsules used for extended life and filtration of highly contaminated solutions
  • Autoclavable capsule filtration device if sterility is required
Filtration of organic solvents and aggressive solutions requires a durable membrane for filtration without sample contamination.
Autoclavable Polycap TF capsule filters feature PTFE membranes with high chemical resistance. Common applications include venting, in-line filtration to sterilize and remove particles from air/gas/liquid streams, filtration to ensure clean inert gases in vial filling lines, and isolation of exhaust from fermenters.
Polycap TF filters support use in the electronics, pharmaceutical, and biopharmaceutical industries, because they are photoresistant and can be used to vent and perform in-line filtration for production systems in biological environments.
Certificaciones: Manufactured according to ISO9001:2008 QMS standards