Order Entry
Acrodisc® Sterile Syringe Filters, Pall® Life Sciences
Acrodisc® Sterile Syringe Filters, Pall® Life Sciences
 :  Cytiva
undefined
Acrodisc® Sterile Syringe Filters, Pall® Life Sciences
 :  Cytiva
  • Catalog No:
  • 28143-298
  • 28143-307
  • Type:
  • Fluorodyne® II
  • Posidyne®
  • Max. operating pressure:
  • 5.4 bar at ambient temperature
  • 5.4 bar at ambient temperature
  • Max. temperature:
  • 60 °C at 4.1 bar
  • 60 °C at 4.1 bar
  • Water flow rate:
  • 130 ml/min. at 2.1 bar (30 psi)
  • 77 ml/min. at 2.1 bar (30 psi)
  •  
    Ø
    Pore size
    Membrane
    Filtration Area
    Supplier No.
  • 28143-298
    4907-N
    25 mm
    0.20 µm
    Fluorodyne® II
    2.8 cm²
    4907
    Paquete de 50
    Paquete de 50 - $7,525.08
    $7,525.08
    Paquete de 50
       $7,525.08
    Product available on GSA Advantage®
    Specifications
    Type
    Fluorodyne® II
    Environmentally Preferable
    Max. operating pressure
    5.4 bar at ambient temperature
    Max. temperature
    60 °C at 4.1 bar
    Water flow rate
    130 ml/min. at 2.1 bar (30 psi)
    Product available on GSA Advantage®
  • 28143-307
    4908-N
    25 mm
    0.20 µm
    Posidyne®
    2.8 cm²
    4908
    Paquete de 50
    Paquete de 50 - $7,525.08
    $7,525.08
    Paquete de 50
       $7,525.08
    Product available on GSA Advantage®
    Specifications
    Type
    Posidyne®
    Environmentally Preferable
    Max. operating pressure
    5.4 bar at ambient temperature
    Max. temperature
    60 °C at 4.1 bar
    Water flow rate
    77 ml/min. at 2.1 bar (30 psi)
    Product available on GSA Advantage®