Whatman™ UniPrep™ Syringeless Filters, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™ UniPrep™
UN513UORG UN113EAQU UN113UORG UN113UGMF UN113UNYL UN113UORG UN113UNYL UN113UAQU UN513UORG UN113ENYL UN113UGMF UN113UAQU UN113ENYL UN113EORG UN513UAQU UN113EAQU UN113EORG UN513UAQU
28297-192EA 267.92 MXN
28297-212 CA28297-188 CA28297-210 CA28297-178 CA28297-212 28297-192 CA28297-192 28297-190 CA28297-174 CA28297-186 CA28297-176 28297-178 28297-188 28297-210 28297-176 28297-186 28297-174 CA28297-190
Whatman™ UniPrep™ Syringeless Filters, Whatman products (Cytiva)
Filtros Filtros sin jeringas
UniPrep™ Syringeless Filters are preassembled filtration devices for the filtration and storage of laboratory samples. These devices are quick and easy to use and feature a plunger, filter, and vial in one unit. They replace syringe-coupled filtration devices with single, disposable units.
  • Integral storage vial saves time and minimizes laboratory waste
  • Built-in glass fiber prefilter means even difficult samples are quick and easy to prepare
  • Choice of membranes for wide sample compatibility
  • Recyclable after use
UniPrep™ devices consist of two parts: a test tube and a filter-plunger. The design incorporates a prefilter and a membrane into the tip of the plunger. When the filter plunger is pressed through the liquid placed in the test tube, positive pressure forces the filtrate up into the reservoir of the filter-plunger.
UniPrep™ devices function in a similar way to the Mini-UniPrep™ device. However, UniPrep™ devices does not contain a septum in the cap and can be used to filter larger volumes (1 to 5 ml).
The UniPrep™ Syringeless Filter is selected based on compatibility with the sample in use. In manual operation, the filter-plunger, after the tip comes in contact with the liquid, is slowly pushed into the test tube until it stops at the bottom. The UniPrep™ device is emptied either by decanting into a sample or autosampler vial or by drawing the filtered sample into a syringe for manual injection into an instrument.