Lambda XLS and Bio Spectrophotometers, PerkinElmer
Proveedor: PerkinElmer
L7110192 L7110194 L7110191 L7110190 L7110185 L7110193 L7110188 L7110192 L7110184 L7110184 L7110187 L7110187 L7110191 L7110189 L7110185 L7110189 L7110186 L7110186 L7110190 L7110188 L7110194 L7110193
97024-668EA 213396.3 MXN
CA97024-668 CA97024-678 CA97024-664 97024-676 97024-666 CA97024-674 CA97024-666 97024-678 97024-668 CA97024-676 CA97024-682 97024-672 97024-662 CA97024-670 97024-674 CA97024-662 CA97024-672 97024-664 97024-680 97024-670 CA97024-680 97024-682
Lambda XLS and Bio Spectrophotometers, PerkinElmer
The simple and elegant LAMBDA™ XLS and Bio UV/Vis spectrophotometers deliver impressive performance in small, easy-to-use packages.
  • Instant answers
  • Large, easy-to-read LCD screen
  • Maintenance-free for continuous uptime
  • Superb stability
  • Rugged design resists solvent spills
No moving parts, virtually no maintenance, and no consumables makes LAMBDA™ XLS and Bio spectrophotometers the perfect tools when the instrument just needs to work, day in and day out
Storage capacity and spectrum bandwidth sizes vary between models.
Low stray light provides exceptional linearity and confidence in your measurements.