Heidolph® MR Hei-End Magnetic Hot Plate Stirrer, Brinkmann®
036110580 036110580CA
89029-840EA 80551.98 MXN
89029-840 CA11009-530
Heidolph® MR Hei-End Magnetic Hot Plate Stirrer, Brinkmann®
Hot plate stirrer features high safety standards with an extra safety control circuit and hot plate cut-out via two independent temperature sensors (not included), as well as an independent safety circuit that switches off the heating if the nominal temperature is exceeded by 10 to 100°C.
Round hot plate is constructed of Silumin aluminum alloy with a ceramic coating. Analog and serial RS-232 interface. (Brinkmann 036110580)
Información para pedidos: Required accessories include a medium temperature sensor (optional) that uses two independent PT 1000 external temperature sensors (not included) to double-check each other (safety circuit). Medium temperature control accuracy of ±0.2K is acheived with external temperature sensor PT 1000 (not included). For more information on sensors and other accessories, contact your VWR representative.