Mini Capsule Filter, 0.2 µm, Sterile, Cytiva (Formerly Pall Lab)
12122 12122
28145-101EA 2585.82 MXN
28145-101 CA28145-101
Mini Capsule Filter, 0.2 µm, Sterile, Cytiva (Formerly Pall Lab)
For final filtration of 1–10L batches with reliable bacterial retention.
  • Two layers
  • Built-in air vent
  • Polycarbonate housing
Incorporates two layers of low protein-binding HT Tuffryn® (hydrophilic polysulfone) membrane: 0.45µm prefilter layer followed by 0.2µm final filter.
Built-in air vent allows elimination of trapped air bubbles. Polycarbonate housing is fitted with 1.27cm (1/2") male, barbed, hose connections for rapid installation with flexible tubing. Biosafe and disposable. Gamma irradiation sterilized.