KIMBLE® Potter-Elvehjem Tissue Grinders with Glass Pestles, DWK Life Sciences
Proveedor: DWK Life Sciences
KONTES®
885500-0024 885500-0019 885500-0022 885500-0023 885500-0021
KT885500-0024EA 3587.1 MXN
KT885500-0021 KT885500-0024 KT885500-0022 KT885500-0023 KT885500-0019
KIMBLE® Potter-Elvehjem Tissue Grinders with Glass Pestles, DWK Life Sciences
Homogenizadores Trituradoras tisulares
For homogenization of connective tissue in muscles, hearts, lungs, etc
  • All-glass construction
  • Designed for a motor drive using a stirrer adapter
  • Clearance between pestles and tubes is 0.1 to 0.15 mm.
  • Replacement components are available and completely interchangeable.
For special clearance pestles, contact your VWR sales representative.
Certificaciones: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.