VWR® Storage/Media Bottles
Proveedor: Avantor
VWR®
VW14396-5000 VW14398-1000 VW14398-2000
89000-240CS 2933.33 MXN
89000-240 89000-242 89012-124
VWR® Storage/Media Bottles
Frascos Frascos de medios
VWR® storage/media bottles are designed with a wide opening (30 mm Int.Ø) and a drip-free polypropylene pour ring.
  • Wide mouth opening with drip-free pour ring
  • Borosilicate glass construction
  • Enhanced graduations
  • Autoclavable
They feature enhanced graduations and marking spots printed in permanent white enamel. Bottles are manufactured from borosilicate glass and can be autoclaved to 180 °C (356 °F). Optional high-temperature cap 89000-946 is autoclavable to 180 °C (356 °F).