Owl™ Aluminum Backed Sequencer System, Models S1S, S2S, S3S and S4S, Thermo Scientific
Proveedor: Thermo Scientific
Owl Series
S3S S4S
27372-796EA 30465.7 MXN
27372-796 27372-788
Owl™ Aluminum Backed Sequencer System, Models S1S, S2S, S3S and S4S, Thermo Scientific
Secuenciadores
Design provides flat, even banding patterns across all lanes and shortens pre-run times.
The upper buffer chamber (UBC) uses an in-line stopcock to drain into a removable container on the back of the unit
Integral side clamps ensure a leakproof seal and even pressure across the gel. Sliding side clamps are mounted on the unit for easy set-up.
Floating, anodized aluminum heat sink moves with the unit as it heats and expands, reducing glass breakage. Heat sink holds plates tightly, providing effective heat dissipation.
A silicone gasket provides a leakproof seal at the UBC. The lower buffer chamber can be removed to drain buffer separately from the UBC. Both chambers are fitted with safety interlocking lids that eliminate the risk of accidental shock.
Información para pedidos: Complete systems include upper buffer chamber with removable drain bottle; removable lower buffer chamber; safety interlocking lids with attached power cords; anodized aluminum heat sink; 1 pair built-in side clamps; 2 blank glass plates; 2 notched glass plates; 2 side spacers, 0.4 mm thick; and 1 adhesive temperature indicator. Also included with gel size 20 x 45 cm system are two 39-tooth sharktooth combs, 0.4 mm thick, and a 20-well standard well comb, 0.4 mm thick. Also included with 35 x 45 cm system are two 78-tooth sharktooth combs, 0.4 mm thick, and a 40-well standard well comb, 0.4 mm thick.