Designed for applications requiring exceptional accuracy, stability, and repeatability, these stirrers are equipped with an exclusive safety system that helps protect both the operator and sample.
- Microprocessor controlled
- Digital display for speed and time
- Chemical-resistant housing
- Now with Best-in-Class, Exclusive 5-Year Warranty
- Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Easy-to-read touch pad controls are located on the front of the unit, with independent LED displays for speed and time
The low-profile design occupies minimal bench space, and permits use in fume hoods. Stirrers are available with ceramic or aluminum top plate material. White reflective ceramic tops are chemical-resistant and easy to clean. Durable aluminum tops will not crack or chip.
Stirrers are equipped with a variety of innovative safety features designed to facilitate efficient operation and workplace safety. Unit housing is constructed of a chemical-resistant polymer. The spill-resistant housing channels fluids away from internal components in the event of a spill. Stirrers have a ramping feature that gradually increases speed to prevent spills, enhance coupling, and provide excellent low end control. In timed mode, an alarm will sound when time reaches zero.
A timer displays elapsed time or a user-defined limit, and will shut off the unit when time reaches zero. The last used settings will be displayed, even after the power has been shut off. Enhanced electronics regulate the stirring, and a powerful continuous duty motor provides reliable and consistent stirring at all speeds.
Certificaciones: RoHS certified. TUV listed. Calibrated Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Información para pedidos: Stirrers include a 234 cm (92") detachable, 3-wire cord and plug. Units are also supplied with a 3.8 cm (11/2") PTFE-coated stir bar. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.