VWR® Spinbar® Magnetic Stir Bars, Polygon without Pivot Ring
Proveedor: VWR
VWR® Spinbar®
37120-6058 37120-6012 37120-6003 37120-6002 37120-6078 37120-6005 37120-6001 37120-6038 37120-6004
58948-988CS 3455.76 MXN
58948-988 58948-986 58948-080 58948-984 58948-985 58948-982 58948-983 58948-980 58948-981
VWR® Spinbar® Magnetic Stir Bars, Polygon without Pivot Ring
Imánes de agitación Barras magnéticas de agitación
Polygon-shaped Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.
  • Suitable for general stirring applications
  • Durable
  • Inert
Polygon-shaped stir bars come without a pivot ring.