VWR® Spinbar® Stir Bars, Octagon
Proveedor: VWR
VWR® Spinbar®
37157-6020 37157-6010 37157-6005 37157-6015
58949-036CS 4008.82 MXN
58949-036 58949-038 58949-034 58949-040
VWR® Spinbar® Stir Bars, Octagon
Imánes de agitación Barras magnéticas de agitación
Convenient blister packs of PTFE resin-coated, Spinbar® magnetic stir bars.
  • PTFE resin coating
  • Alnico magnet
  • Most commonly used shape
Alnico magnets encapsulated in FDA grade Teflon® PTFE are durable and inert.