VWR® Spinbar® Micro Stir Bars
Proveedor: Avantor
VWR® Spinbar®
37119-6011 37119-6183 37119-6003 37119-6001 37119-6010 37119-6008 37119-6005 37119-6004 37119-6007 37119-6009 37119-6002
76001-878CS 2922.15 MXN
76001-878 76001-876 58948-976 76001-874 76001-872 58948-377 58948-411 58948-400 58948-375 58948-397 58948-353
VWR® Spinbar® Micro Stir Bars
Imánes de agitación Barras magnéticas de agitación
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes, and gradient makers.
  • Particularly useful for environmental testing
  • Valuable for preparing and evaluating small sample volumes
  • Teflon® PTFE encapsulated
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.