VWR® Spinvane® Stir Bars for Test Tubes, Micro Vials, and Conical Bottom Centrifuge Tubes
Proveedor: VWR
VWR® Spinvane®
37134-9001 37134-6001 37136-6001 37135-6001 37137-6001
55999-000PK 8718.84 MXN
55999-000 58949-274 58949-276 58949-270 58949-272
VWR® Spinvane® Stir Bars for Test Tubes, Micro Vials, and Conical Bottom Centrifuge Tubes
Imánes de agitación Barras magnéticas de agitación
Solid Teflon® vane has a cross-mounted, molded-in magnet that can be trimmed to fit if needed without affecting the magnet.
  • Half-round shape is designed for 16 mm O.D. test tubes
  • Half-round tapered shape is designed for conical-bottom centrifuge tubes
  • Triangular shape is designed for small conical-shaped tubes and micro vials
  • Autoclavable
Each size is manufactured for a specific size tube; can be trimmed to fit if needed without affecting the magnet.