VWR® Spinplus® Magnetic Stir Bars
Proveedor: VWR
VWR® Spinplus®
37144-6038 37188-6000 37144-6114 37144-6112 37144-6100 37144-6034
58947-826CS 5869.71 MXN
58947-826 58947-828 58947-830 58947-820 58947-822 58947-824
VWR® Spinplus® Magnetic Stir Bars
Imánes de agitación Barras magnéticas de agitación
Spinplus® magnetic stirring bars add speed and efficiency to mixing operations.
  • The “+” shape creates a deep vortex and provides stable, quiet operation
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating is durable and inert.
Información para pedidos: Order each size separately or choose a five-piece assortment (one of each size).