VWR® Polygon Spinbar Magnetic Stirring Bars
Proveedor: VWR
VWR® Spinbar®
F37122-0070 F37122-0025 F37122-0040 F37122-0045 F37122-0060 F37122-0050 F37122-0012 F37122-0020 F37122-0035
74950-288EA 147.89 MXN
74950-288 74950-300 74950-286 74950-298 74950-284 74950-296 74950-294 74950-292 74950-290
VWR® Polygon Spinbar Magnetic Stirring Bars
Imánes de agitación Barras magnéticas de agitación
These VWR® stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars
  • Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
  • Marring of plastic containers is diminished
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® brand PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe