Stainless Steel Worktables, Rounded Edge, Bandy
Proveedor: Bandy
TBL0704-2424 TBL0706-2484 TBL0706-3072 TBL0704-2472 TBL0706-2424 TBL0704-3048 TBL0704-3036 TBL0706-3660 TBL0704-3060 TBL0704-2448 TBL0704-3648 TBL0704-3030 TBL0706-2472 TBL0704-3660 TBL0704-2436 TBL0704-2430 TBL0706-3036 TBL0706-2436 TBL0706-2448 TBL0706-3060 TBL0704-3684 TBL0704-3072 TBL0706-2460 TBL0706-3684 TBL0704-3672 TBL0704-2460 TBL0706-3084 TBL0704-2484 TBL7001-3048 TBL0706-3030 TBL0704-3636 TBL0706-3648 TBL0706-3048 TBL0704-3084
97001-568EA 25781.6 MXN
97001-420 97001-442 97001-520 97001-542 97001-422 97001-488 97001-466 97001-522 97001-424 97001-568 97001-502 97001-546 97001-480 97001-460 97001-482 97001-560 97001-582 97001-440 97001-562 97001-538 97001-530 97001-454 97001-554 97001-478 97001-578 97001-512 97001-536 97001-470 97001-570 97001-494 97001-550 98000-610 97001-506 97001-508
Stainless Steel Worktables, Rounded Edge, Bandy
Mobiliario Mesas
Worktables are constructed of 16-gauge, Type 304 stainless steel by providing a sturdy work area in the cleanroom.
  • 1⁵/₈" Round tube legs with socket connectors
  • Adjustable leveling feet
  • Medium duty work bench
Tabletop has rounded front and rear edges, and the sides are turned down on a 90° angle for easy alignment with other equipment. Table legs have a 4.1 cm (1⁵/₈") diameter and a 2.5 cm (1") height adjustment.
Working height: 76.2 cm (30")
Información para pedidos: Shipped unassembled. Plastic leveling feet included. Available with or without backsplash or 18-gauge stainless steel undershelf.