Paddles, Chain, Talboys
156XL 157XL 155L 155XL 157 155 156 157L 156L
14010-016EA 1248.29 MXN
58950-700 58950-704 14010-020 58950-702 58950-692 14010-016 58950-696 14010-018 58950-694
Paddles, Chain, Talboys
These paddles are useful for mixing the contents of narrow-necked containers and fit openings as small as 16mm (5/8") wide
Both chain and shaft are made of Type 303 stainless steel. Swing diameter of the chain is approximately 76mm (3").
Chain paddles also can stir both low- and high-viscosity liquids, and blend suspensions.