VWR® Spinbar® Micro Stir Bars, Color-Coded, Assortment
Proveedor: VWR
VWR® Spinbar®
37186-6000
58948-076CS 12278.11 MXN
58948-076
VWR® Spinbar® Micro Stir Bars, Color-Coded, Assortment
Imánes de agitación Barras magnéticas de agitación
Assortment of 12 micro-magnetic, PTFE resin-coated stir bars color-coded in blue, yellow, and red.
  • Color-coded
  • Magnet encapsulated in PTFE coating
  • Durable
  • Inert
Packaged in a plastic, compartmentalized Spinbox™.
Información para pedidos: Includes one of each color in four different sizes: 8x1.5mm (5/16x1/16"), 5x2mm (3/16x1/8"), 7x2mm (5/16x1/8"), and 10x3mm (7/16x3/16").