VWR® Spinbar® Magnetic Stir Bars, Egg-Shaped
Proveedor: VWR
VWR® Spinbar®
37130-6004 37130-6002 37130-6006 37130-6038 37130-6008 37130-6007 37130-6058 37130-6001 37130-6005 37130-6003
58949-006CS 4802.49 MXN
58949-006 58949-008 58949-002 58949-200 58949-016 58949-196 58949-010 58949-012 58949-243 58949-210
VWR® Spinbar® Magnetic Stir Bars, Egg-Shaped
Imánes de agitación Barras magnéticas de agitación
These egg-shaped stirring bars, designed especially for use in round-bottom vessels, spin unrestricted by the vessel contour.
  • Excellent for round-bottom flasks
  • Provide their own pivot point without a separate ring or abrupt change in contour
  • Offers minimal contact when used in plastic containers
The magnet itself is egg-shaped with a Teflon® PTFE coating, except the three smallest sizes have a cylindrical magnet in an egg-shaped Teflon® PTFE coating. Autoclavable.