Sure-Grip® EX Safety Cabinets for Flammable Materials, Justrite®
Proveedor: Justrite
Sure-Grip®
890420 890400 899000 891300 8945253 893020 894580 891200 894500 8930253 8945053 894520 891700 891320 892300 891720 899020 896000 8930053 896020 891500 893300 893400 896080 891520 891220 892320 899080 890500
97001-366EA 47788.6 MXN
97049-640 97001-366 97001-400 97006-122 97001-368 97001-248 97006-142 97006-120 97006-140 97033-754 97001-384 97001-362 97001-342 97001-364 97006-118 97006-116 97049-636 97006-114 97001-410 97006-112 97001-238 97006-130 97001-390 97001-372 97001-352 97001-210 76018-056 97006-128 97006-126
Sure-Grip® EX Safety Cabinets for Flammable Materials, Justrite®
Mobiliario Armarios de almacenamiento
Countertop and Compac cabinets provide safe storage when space is limited
  • Fire-resistant, welded 18-gauge steel
  • Double-wall design features a 3.8 cm (1/2”) insulating air space
  • Built-in grounding connector prevents static buildup
  • Adjustable shelves
Fire-resistant, welded 18-gauge steel cabinets store flammable liquids and waste safely and securely, protecting personnel and facilities. The double-wall design features a 3.8cm (11/2") insulating air space. A 3-point stainless steel bullet latching system further reduces the risk of fire. Exclusive features include U-Loc™ handles that include two keys and accept padlocks, Haz-Alert™ reflective exterior labeling, and galvanized steel SpillSlope™ shelves that direct all spills to the 5.1cm (2") leakproof sump. Adjustable shelves provide a 159kg (350lb.) safe allowable load. Built-in grounding connector prevents static buildup. Four adjustable leveling feet ensure a wobble-free base.
Undercounter cabinets offer quick access at workstations and are sized and fitted with a recessed kickplate to mount flush with existing cabinetry. Wall mount cabinets (hardware included) are ideal for small container storage and can be positioned throughout the workplace. Piggyback cabinets are designed to mount atop standard 114L (30gal.) and 117L (45gal.) cabinets, but can also be used independently as benchtop units. Flammable waste cabinets are designed for temporary collection and segregation of waste liquids.
Certificaciones: All cabinets are OSHA compliant, meet NFPA Code 30, and are independently tested and FM approved.
Información para pedidos: All cabinets include manufacturer's 10-year warranty. See 97001-314 series for extra shelves and other accessories.