Sure-Grip® EX Drum Storage Cabinets, Justrite®
Proveedor: Justrite
Sure-Grip®
8962053 896200 899300 8991053 896220 899320 899100
97001-408EA 68120.14 MXN
97006-136 97001-376 97001-414 97001-404 97001-416 97001-408 97001-374
Sure-Grip® EX Drum Storage Cabinets, Justrite®
Mobiliario Armarios de almacenamiento
Fire-resistant steel cabinets store flammable liquids and waste safely and securely, protecting both personnel and facilities
  • Exclusive Sure-Grip® EX features include U-Loc™ handles that include two keys and accept padlocks
  • Haz-Alert™ reflective exterior labeling for easy identification in fire conditions or power outages
  • Galvanized steel SpillSlope™ shelves safely direct all spills to the 5.1cm (2") leakproof sump
Horizontal single drum cabinets are designed for drums being used for gravity flow dispensing. They provide space for a drip can and are equipped with a cradle track to assist in guiding drums into position. Vertical single and double drum cabinets include one half-depth storage shelf, yet provide enough space to accommodate a drum equipped with a lower height pump or funnel to allow the cabinet to be used as a pumping station or for waste accumulation. These cabinets include a removable steel drum support in the unit base to keep all contents raised off of the sump. Flammable waste drum cabinets allow for the temporary accumulation and storage of waste liquids prior to disposal.
Cabinets are constructed of sturdy 18-gauge steel and are welded for strength, a long service life, and increased fire protection. The double-wall design features a 3.8cm (11/2") insulating air space. A three-point stainless steel bullet latching system further reduces the risk of fire. All cabinets have two doors that are either manual- or self-closing.
Certificaciones: All cabinets are OSHA compliant, meet NFPA Code 30, and are independently tested and FM approved.
Información para pedidos: All cabinets are supplied with manufacturer's 10-year warranty. For cabinets with roller assemblies, contact your Avantor sales representative.