Axygen® PCR Tube Storage Racks, Corning
Proveedor: Corning
Axygen®
R-96-PCR-FSP R-96-PCR-FY
80086-082CS 4500 MXN
80086-082 80086-074
Axygen® PCR Tube Storage Racks, Corning
Gradillas Tube Racks
PP with lid.
  • Fluorescent color base with translucent lid
  • Durable, chemically inert, and can withstand extended duration at sub-zero temperatures
Designed to hold 0.2, and 0.5 ml PCR tubes.
Información para pedidos: Assorted pack includes one each of blue, green, violet, red and yellow.