Continuous Agitation Mixer, Arrow
THE AGITATOR
59025-132EA 32439.53 MXN
59025-132
Continuous Agitation Mixer, Arrow
This powerful mixer is designed for applications requiring high agitation
Mixer features variable stirrer speed control and reversing time cycle which permits selection of extremely dynamic and violent mixing action without formation of air bubbles. Direct-drive, permanent magnet DC motor is totally enclosed, non-ventilated, and specially designed to handle viscous fluids. Unit can be used as a regular mixer by operating it in one direction. Maximum volume that stirrer can handle is dependent on viscosity, shaft length, and speed at which liquid is stirred.
Instant forward-and-reverse action provides powerful mixing action that keeps matter in suspension without vortexing.
Información para pedidos: Complete with 25.4L x 1.4dia. cm (10 x 9/16") mounting rod, chemical-resistant stainless steel paddle assembly, three-wire cord and plug, and built-in circuit breaker. Shaft measures 30.5L x 1dia. cm (12 x 3/8") with a 6.4cm (21/2") diameter propeller.