Provides accurate and reliable titration results with the convenience of a large digital display, eliminating tedious meniscus readings.
- Safer and more accurate than glass
- Air purge and safety shutdown
- Recirculating valve system
- 360° swivel neck
The buret attaches directly to the reagent bottle, eliminating the hazards of pouring reagents
Tight control over reagent flow is maintained by easy-to-turn wheels on the side of the unit.
Multiple titrations can be stored in memory with a maximum of 500 values. Using the optional interface and software package, stored values can be downloaded to a PC.
Designed to hold 50 mL of reagent so refilling during a titration does not compromise accuracy. With the unit's unique recirculating valves, air is purged from the system without loss of reagents. The valves can also be rotated to the safety position to prevent accidental discharge when the buret is not in use.
Información para pedidos: Burets include a rechargeable NiCad battery, AC adapter/recharger, and adapters to fit reagent bottles with 33mm, 38mm, [ST] 24/40, and [ST] 29/42 neck sizes. Burets 82017-946 and -948 also feature a fine volume control knob. Bottle is not included.