PYREX® Burets, Class A, Colored Graduations, Fluoropolymer Resin Stopcock, Corning
Proveedor: Corning
PYREX®
2103-50 2103-10 2103-100 2103-25
17452-404CS 3000 MXN
17452-404 17452-426 17452-368 17452-380
PYREX® Burets, Class A, Colored Graduations, Fluoropolymer Resin Stopcock, Corning
Buretas
Borosilicate glass, class A.
  • Red colored graduations feature permanent markings, fine clear lines
  • Large easy to read numbers
  • Smooth micro-finish stopcock assures leak-resistant performance
Burette tips are carefully drawn from precision bore tubing to ensure proper drainage rates.
Certificaciones: Capacity tolerance is established by ASTM E287 and calibrated in accordance with ASTM E542.
Información de suministro: Burets are shipped complete with PTFE stopcock and dust cover.